missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC170 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 590.10€
Specifications
| Antigen | CCDC170 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635698
|
Novus Biologicals
NBP2-48579-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627897
|
Novus Biologicals
NBP2-48579 |
0.1 mL |
624.00€ 590.10€ / 0.10mL Save 33.90€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCDC170 Polyclonal antibody specifically detects CCDC170 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CCDC170 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bA282P11.1, C6orf97, chromosome 6 open reading frame 97, coiled-coil domain containing 170, FLJ23305, hypothetical protein LOC80129 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ETINVHEMEAKASRETIMRLASEVNREQKKAASCTEEKEKLNQDLLSAVEAKEALEREVKIFQERLLAGQQVWDASKQEVSLLKKSSSELEKSLKASQDAVT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 80129 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title