missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC109A Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33367-100ul
This item is not returnable.
View return policy
Description
CCDC109A Monoclonal antibody specifically detects CCDC109A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CCDC109A | |
| Monoclonal | |
| Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| C10orf42, coiled-coil domain containing 109A, coiled-coil domain-containing protein 109A, FLJ46135 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 51-170 of human CCDC109A (NP_612366.1).,, Sequence:, VHQRIASWQNLGAVYCSTVVPSDDVTVVYQNGLPVISVRLPSRRERCQFTLKPISDSVGVFLRQLQEEDRGIDRVAIYSPDGVRVAASTGIDLLLLDDFKLVINDLTYHVRPPKRDLLSH | |
| 100 μL | |
| Cardiovascular Biology, Endocrinology | |
| 90550 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction