missing translation for 'onlineSavingsMsg'
Learn More

Ccd1/DIXDC1 Antibody, Novus Biologicals™

Product Code. 18289712 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18289712 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18289712 Supplier Novus Biologicals Supplier No. NBP146147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Ccd1/DIXDC1 Polyclonal specifically detects Ccd1/DIXDC1 in Human samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ccd1/DIXDC1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001033043.1
Gene Alias CCD1, Coiled-coil protein DIX1, Coiled-coil-DIX1, DIX domain containing 1, DIX domain-containing protein 1, dixin, KIAA1735coiled-coil-DIX1
Gene Symbols DIXDC1
Host Species Goat
Immunogen Peptide with sequence C-DVSYHQVDLERE corresponding to internal region according to NP_219493.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 85458
Test Specificity This antibody is expected to recognize both isoforms (NP_001033043.1; NP_219493.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.