missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | Caveolin-2 |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 5-10 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692866
|
Novus Biologicals
NBP2-99542-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677655
|
Novus Biologicals
NBP2-99542-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caveolin-2 Polyclonal antibody specifically detects Caveolin-2 in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Caveolin-2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Caveolae Markers, Membrane Trafficking and Chaperones, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 858 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:1000-1:5000, Immunoprecipitation 5-10 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CAV, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, MGC12294 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Caveolin-2 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title