missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav3.2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€ - 507.00€
Specifica
| Antigen | Cav3.2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
18626947
|
Novus Biologicals
NBP2-41303-0.025mg |
0.025 mg |
178.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166149
|
Novus Biologicals
NBP2-41303 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Cav3.2 Polyclonal specifically detects Cav3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifica
| Cav3.2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 8912 | |
| Antibody was raised against a 16 amino acid peptide near the center of human CACNA1H. The immunogen is located within amino acids 1010 - 1060 of CACNA1H. Amino Acid Squence: AEGDANRSDTDEDKTS | |
| Primary | |
| Multiple isoforms of CACNA1H are known to exist. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| CACNA1HB, calcium channel, voltage-dependent, T type, alpha 1H subunit, calcium channel, voltage-dependent, T type, alpha 1Hb subunit, Cav3.2, ECA6, EIG6, FLJ90484, Low-voltage-activated calcium channel alpha1 3.2 subunit, low-voltage-activated calcium channel alpha13.2 subunit, voltage dependent t-type calcium channel alpha-1H subunit, voltage-dependent T-type calcium channel subunit alpha-1H, voltage-gated calcium channel alpha subunit Cav3.2, voltage-gated calcium channel alpha subunit CavT.2, Voltage-gated calcium channel subunit alpha Cav3.2 | |
| CACNA1H | |
| IgG | |
| Affinity Purified | |
| 260 kDa |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto