missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cav1.3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92203-0.02ml
This item is not returnable.
View return policy
Description
Cav1.3 Polyclonal antibody specifically detects Cav1.3 in Mouse samples. It is validated for Western Blot
Specifications
| Cav1.3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1882-2181 of human CACNA1D (NP_000711.1). RQNYGYYSRYPGRNIDSERPRGYHHPQGFLEDDDSPVCYDSRRSPRRRLLPPTPASHRRSSFNFECLRRQSSQEEVPSSPIFPHRTALPLHLMQQQIMAVAGLDSSKAQKYSPSHSTRSWATPPATPPYRDWTPCYTPLIQVEQSEALDQVNGSLPSLHRSSWYTDEPDISYRTFTPASLTVPSSFRNKNSDKQRSADSLVEAVLISEGLGRYARDPKFVSATKHEIADACDLTIDEMESAASTLLNGNVRPRANGDVGPLSHRQDYELQDFGPGYSDEEPDPGRDEEDLADEMICITTL | |
| 0.02 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 776 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction