missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Antibody (C2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43441
This item is not returnable.
View return policy
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Rat samples. It is validated for Western Blot
Specifications
| Cathepsin S | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| Immunized with recombinant Rat Cathepsin S (Uniprot#: Q02765; Leu113∽Ile330) | |
| 100 μL | |
| Immunology | |
| 1520 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| C2 | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction