Learn More
Cathepsin S Antibody (9B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26639-50ul
 View more versions of this product
Product Code. 30043754
  • 343.00€ / 50µL
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cathepsin S
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
9B10
ELISA, Immunohistochemistry 1:50-1:200
cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
A synthesized peptide derived from Human Cathepsin S [UniProt P25774]
50 μL
Immunology
1520
Store at -20 to -70C. Avoid freeze-thaw cycles.