Learn More
Cathepsin S Antibody (9B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26639-100ul
This item is not returnable.
View return policy
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| Cathepsin S | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 9B10 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| A synthesized peptide derived from Human Cathepsin S [UniProt P25774] | |
| 100 μL | |
| Immunology | |
| 1520 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |