missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Catalase (Bovine), 2X

Product Code. 16282240
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16282240 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16282240 Supplier Abnova™ Supplier No. P5220.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P00432
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 531682
Molecular Weight (g/mol) 240kDa
Name Catalase (Bovine), 2x
Preparation Method Native protein purified from Bovine Liver
Quantity 10 mL
Storage Requirements Store at 4°C. Do not freeze. This product is stable for at least 3 months.
Regulatory Status RUO
Gene Alias MGC128112
Common Name CAT
Gene Symbol CAT
Assays Catalase Assay; Free radical research; Deoderization; Decomposition of residual hydrogen peroxide after bleaching woven and knitted cotton fabrics before drying; Cysteamine determination; Gluconic and glycolic acid production; Cleaning silicon and semiconductor plates.
Biological Activity MinimμM of 30,000 units/ml. One Unit decomposes one micromole of hydrogen peroxide per minute at 25°C, pH 7.0.
Species Bovine
Protein Tag None
Expression System Native protein purified from Bovine Liver
Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • 10ML Catalase, 2x
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium phosphate monobasic, potassium phosphate dibasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.