missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 2 Monoclonal Antibody (691233)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561786
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Caspase 2 gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.
Specifications
| Caspase 2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P42575 | |
| CASP2 | |
| E. coli-derived recombinant human Caspase-2, Gly170-Asp333 & Ala348-Tyr452. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 691233 | |
| Unconjugated | |
| CASP2 | |
| CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction