missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol EP Impurity D, Mikromol™

Product Code. 30051066
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30051066 100 mg 100mg
30050944 25 mg 25mg
30462461 4 x 25 mg -
3 options
This item is not returnable. View return policy

Product Code. 30051066

Brand: Mikromol MM0291.13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carvedilol EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(9H-Carbazol-4-yloxy)-3-[4-[2-hydroxy-3-[(2-(2-methoxyphenoxy)ethyl)amino]propoxy]-9H-carbazol-9-yl]propan-2-ol
CAS 1391052-16-6
Analyte or Component Names 1-(9H-Carbazol-4-yloxy)-3-[4-[2-hydroxy-3-[(2-(2-methoxyphenoxy)ethyl)amino]propoxy]-9H-carbazol-9-yl]propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C39 H39 N3 O6
Certifications/Compliance ISO 9001
Synonym 1-(9H-Carbazol-4-yloxy)-3-[4-[2-hydroxy-3-[[2-(2-methoxyphenoxy)ethyl]amino]propoxy]-9H-carbazol-9-yl]propan-2-ol, 9H-Carbazole-9-ethanol, alpha-[(9H-carbazol-4-yloxy)methyl]-4-[2-hydroxy-3-[[2-(2-methoxyphenoxy)ethyl]amino]propoxy]-, Carvedilol Impurity D
InChI Formula InChI=1S/C39H39N3O6/c1-45-34-16-6-7-17-35(34)46-21-20-40-22-26(43)24-47-37-19-9-15-33-39(37)29-11-3-5-14-32(29)42(33)23-27(44)25-48-36-18-8-13-31-38(36)28-10-2-4-12-30(28)41-31/h2-19,26-27,40-41,43-44H,20-25H2,1H3
SMILES COc1ccccc1OCCNCC(O)COc2cccc3c2c4ccccc4n3CC(O)COc5cccc6[nH]c7ccccc7c56
IUPAC Name 1-[9-[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]carbazol-4-yl]oxy-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
Molecular Weight (g/mol) 645.74
Quantity 100 mg
Formula Weight 645.2839 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.