missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARS Antibody (CL2302) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44480
This item is not returnable.
View return policy
Description
CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| CARS | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246 | |
| This antibody was generated using a recombinant protein sequence of A8MVQ3, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 833 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL2302 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction