missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cardiotrophin-1/CT-1 Antibody (89215) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2601P
This item is not returnable.
View return policy
Description
Cardiotrophin-1/CT-1 Monoclonal antibody specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot,Neutralization
Specifications
| Cardiotrophin-1/CT-1 | |
| Monoclonal | |
| PE | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1 | |
| E. coli-derived recombinant human Cardiotrophin-1/CT-1, Ser2-Ala201, Accession # Q16619 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Protein Kinase | |
| 1489 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Neutralization | |
| 89215 | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction