missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD11/CARMA1 Antibody (SR1136), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22214-100ul
This item is not returnable.
View return policy
Description
CARD11/CARMA1 Monoclonal antibody specifically detects CARD11/CARMA1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CARD11/CARMA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1136 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| BIMP3card-maguk protein 1, CARD-containing MAGUK protein 1, Carma 1, CARMA1bcl10-interacting maguk protein 3, caspase recruitment domain family, member 11, caspase recruitment domain-containing protein 11, MGC133069 | |
| A synthesized peptide derived from human CARD11/CARMA1 (Uniprot #: Q9BXL7) | |
| 100 μg | |
| Apoptosis | |
| 84433 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction