missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbromal, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Carbromal
Specifications
Specifications
| Chemical Name or Material | Carbromal |
| CAS | 77-65-6 |
| Analyte or Component Names | Carbromal |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C7 H13 Br N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Carbromal, Butanamide, N-(aminocarbonyl)-2-bromo-2-ethyl-, Carbromal (6CI), Urea, (2-bromo-2-ethylbutyryl)- (8CI), 2-Bromo-2-ethylbutyrylurea, Adalin, Adisomnol, Bromacetocarbamide, Bromadal, Bromdiethylacetylurea, Bromodiethylacetylcarbamide, Diacid, Dormiturin, Hoggar, Karbromal, Kartryl, NSC 49191, NSC 9916, Nenesin, Nyctal, Parkosed, Pelidorm, Planadalin, Tildin, Uradal, (alpha-Bromo-alpha-ethylbutyryl)carbamide, (alpha-Bromo-alpha-ethylbutyryl)urea, alpha-Ethyl-alpha-bromobutyrylurea |
| InChI Formula | InChI=1S/C7H13BrN2O2/c1-3-7(8,4-2)5(11)10-6(9)12/h3-4H2,1-2H3,(H3,9,10,11,12) |
| SMILES | CCC(Br)(CC)C(=O)NC(=O)N |
| IUPAC Name | 2-bromo-N-carbamoyl-2-ethylbutanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction