Learn More
Invitrogen™ Carbonic Anhydrase XII Monoclonal Antibody (336103)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561142
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Carbonic Anhydrase XII is a protein coding gene.
Specifications
| Carbonic Anhydrase XII | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8CI85 | |
| Car12 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase XII/CA12, Ala25-Ser301. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 336103 | |
| Unconjugated | |
| Car12 | |
| 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 76459 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |