missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XII/CA12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57791-25ul
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XII/CA12 Polyclonal specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Carbonic Anhydrase XII/CA12 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| CA12 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NKQFLLTNNGHSVKLNLPSDMHIQGLQSRYSATQLHLHWGNPNDPHGSEHTVSGQHFAAEL | |
| 25 μL | |
| Lipid and Metabolism | |
| 771 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction