missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase X Monoclonal Antibody (862319)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561116
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
Specifications
| Carbonic Anhydrase X | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NS85 | |
| CA10 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase X/CA10, Met1-Asn300. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 862319 | |
| Unconjugated | |
| CA10 | |
| 2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction