missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VIII/CA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99319-50ul
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VIII/CA8 Polyclonal antibody specifically detects Carbonic Anhydrase VIII/CA8 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Carbonic Anhydrase VIII/CA8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 | |
| Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase VIII/CA8 (Accession#: NP_004047.3; Met 1-Gln 290) | |
| 50 μg | |
| Lipid and Metabolism | |
| 767 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction