missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Carbonic Anhydrase VIII/CA8 Antibody (CA8/8605R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26749-100ug
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VIII/CA8 Monoclonal antibody specifically detects Carbonic Anhydrase VIII/CA8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Carbonic Anhydrase VIII/CA8 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 | |
| Recombinant full-length human Carbonic Anhydrase VIII/CA8 protein | |
| 100 μg | |
| Lipid and Metabolism | |
| 767 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CA8/8605R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction