missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VB/CA5B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | Carbonic Anhydrase VB/CA5B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608365
|
Novus Biologicals
NBP2-99972-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621596
|
Novus Biologicals
NBP2-99972-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbonic Anhydrase VB/CA5B Polyclonal antibody specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| Carbonic Anhydrase VB/CA5B | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 11238 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 | |
| Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase VB/CA5B (Uniprot#: Q9Y2D0; Cys34-Pro317) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title