missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX Mouse anti-Human, BV 421, Clone: M75, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 753821
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IX Monoclonal antibody specifically detects Carbonic Anhydrase IX in Human samples. It is validated for Flow Cytometry
Specifications
| Carbonic Anhydrase IX | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
| IgG2b |
| Flow Cytometry | |
| M75 | |
| Brilliant Violet 421 | |
| CA-IX; CAIX; CA9; CAH9; Carbonic anhydrase 9 | |
| MaTu-infected HeLa Cells | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction