missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV/CA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88225
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IV/CA4 Polyclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Carbonic Anhydrase IV/CA4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CA4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GTQVNEGFQPLVEALSNIPKPEMSTTMAESSLLDLLPKEEKLRHYFRYLGSLTTPTCDEKVVWTVFREPIQLHREQILAFSQKLYYDKEQTVSMKDNVRPLQQLGQRTVIKSGA | |
| 0.1 mL | |
| Lipid and Metabolism, Vision | |
| 762 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur