missing translation for 'onlineSavingsMsg'
Learn More

Phoxim 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30328473
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30328473

Brand: Dr. Ehrenstorfer DREA16150000AC1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phoxim
CAS 14816-18-3
Analyte or Component Names Phoxim
Shipping Condition +20°C
Molecular Formula C12 H15 N2 O3 P S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphorothioic acid, O-[(cyanophenylmethylene)azanyl] O,O-diethyl ester (ACI), 3,5-Dioxa-6-aza-4-phosphaoct-6-ene-8-nitrile, 4-ethoxy-7-phenyl-, 4-sulfide (9CI), Glyoxylonitrile, phenyl-, oxime O,O-diethyl phosphorothioate (8CI), Phosphorothioic acid, O,O-diethyl ester, O-[(α-cyanobenzylidene)amino] deriv. (8CI), B 77488, Basileum SI, Basileum SI 84, Basileum SI 84EC, BAY 5621, BAY 77488, Bayer 5621, Bayer 77488, Baythion, Benzoyl cyanide O-(diethoxyphosphinothioyl)oxime, Folaton, Jiubaojing, O,O-Diethyl O-(α-cyanobenzylideneamino) phosphorothioate, Phenylglyoxylonitrile oxime O,O-diethyl phosphorothioate, Sebacil, Tomiguard PE 50, Toyothion, Valexon, Volaton, Volaton bjdse 20, Xylasan VL
InChI Formula InChI=1S/C12H15N2O3PS/c1-3-15-18(19,16-4-2)17-14-12(10-13)11-8-6-5-7-9-11/h5-9H,3-4H2,1-2H3/b14-12-
SMILES CCOP(=S)(OCC)O\N=C(\C#N)/c1ccccc1
IUPAC Name (E)-N-diethoxyphosphinothioyloxybenzenecarboximidoyl cyanide
Solvent or Matrix Acetone
Molecular Weight (g/mol) 298.3
Quantity 1 mL
Formula Weight 298.0541 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.