Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Click to view available options
For Use With (Equipment):
Smart 321 Fume Hood
Smart 391 Fume Hood
Smart 392 Fume Hood
Smart 481 Fume Hood
Smart 483 Fume Hood
Smart 633 Fume Hood
Smart 714 Fume Hood
Förpackningsstorlek
Each
Beskrivelse
- Made of phenolic resin with ergonomic arm for a comfortable working position
- High chemical and mechanical resistance
- Ideal for precision weighing operations
Tekniske data
Tekniske data
| For Use With (Equipment) | Smart 481 Fume Hood |
| Includes | Spill Tray |
| Depth (Metric) | 545 mm |
| Width (Metric) | 1190 mm |
| Product Type | Work Surface |
| Material | Phenolic Resin |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion