Learn More
Erlab™ Captair by Erlab™ H402 Version 03 Filtration Columns
Product Code. p-8011757
Click to view available options
Filter Type:
BE Filter
Unit Size:
Each
Product Code. 12157720
Brand: Erlab™ G0103/BE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ H402 Version 03 Filtration Columns are for storing organic vapors and acids
  • Horizontal filtration column
  • HEPA H14 filter
  • Handlings in an ultra clean environment
  • Serial prefilter(optimizes the HEPA H14 filter lifetime)
  • Optional carbon filtration = clean the air polluted by ambient air VOCs
Specifications
Product Type Filtration Column
Filter Type BE Filter
For Use With (Application) For storing organic vapors and acids