Learn More
Erlab™ Captair by Erlab™ H402 Version 03 Filtration Columns
Product Code. 12157720 Shop All Erlab Products
Change view
Click to view available options
Filter Type:
BE Filter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type Unit Size
12157720 BE Filter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12157720 Supplier Erlab™ Supplier No. G0103/BE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ H402 Version 03 Filtration Columns are for storing organic vapors and acids
  • Horizontal filtration column
  • HEPA H14 filter
  • Handlings in an ultra clean environment
  • Serial prefilter(optimizes the HEPA H14 filter lifetime)
  • Optional carbon filtration = clean the air polluted by ambient air VOCs
Specifications
Product Type Filtration Column
Filter Type BE Filter
For Use With (Application) For storing organic vapors and acids