missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R2/CB2/CNR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
213.00€
Specifications
| Antigen | Cannabinoid R2/CB2/CNR2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:250 - 1:2500, Flow Cytometry 1:50, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Cannabinoid R2/CB2/CNR2 Polyclonal specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Cannabinoid R2/CB2/CNR2 | |
| Western Blot 1:250 - 1:2500, Flow Cytometry 1:50, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| Polyclonal | |
| Rabbit | |
| GPCR, Neuroscience, Vision | |
| PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide | |
| 1269 | |
| A fusion protein containing the first 33 amino acid residues from human CB2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| cannabinoid receptor 2 (macrophage), CB-2, CB2cannabinoid receptor 2, CX5testis-dominant CNR2 isoform CB2, hCB2 | |
| CNR2 | |
| IgG | |
| Affinity Purified | |
| Cannabinoid Receptor 2 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title