missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cannabinoid R1/CB1/CNR1 Antibody (GFB-024) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28839-1mg
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Cannabinoid R1/CB1/CNR1 | |
| Monoclonal | |
| Unconjugated | |
| Q96F85 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| GFB-024 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
| CB1/ CNR1 | |
| 1 mg | |
| Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
| 1268 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction