Learn More
ACROBiosystems Canine IL-3 R alpha / CD123 Protein, His Tag
Brand:  ACROBiosystems ILA-C52H3-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17206270
  • 3357.52€ / 1mg
Estimated Shipment: 07-08-2024
Description
Interleukin 3 receptor alpha (low affinity) (IL3RA), also known as CD123 (Cluster of Differentiation 123) is a 70-kD glycoprotein member of the hematopoietin receptor superfamily. This protein associates with a beta subunit common to the receptors for IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to form a high-affinity receptor for IL-3. The interleukin-3 receptor α chain (CD123) has been identified as a potential immunotherapeutic target because it is overexpressed in AML compared with normal hematopoietic stem cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa.
Specifications
Specifications
XP_038305195.1
1 mg
Ser 33 - Asp 315
IL3R,IL3RA,IL-3 Ra,IL-3 R-alpha,IL3RAY,IL3RX,IL3RY,CD123 antigen,CD123,hIL3Ra,hIL-3 Ra,MGC34174,IL-3 R alpha
Unconjugated
His Tag
Powder
34.9 kDa
Canine IL-3 R alpha, His Tag (ILA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Asp 315 (Accession # XP_038305195.1).
-20°C
IL-3 R alpha
Canine
HEK293
Bio-Markers & CD Antigens