missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Mouse anti-Human, Clone: CALB2/2602, Novus Biologicals™
Mouse Monoclonal Antibody
507.00€
Specifications
| Antigen | Calretinin |
|---|---|
| Clone | CALB2/2602 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
Calretinin Monoclonal specifically detects Calretinin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Calretinin | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 794 | |
| IgG1 κ | |
| Protein A purified |
| CALB2/2602 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| Recombinant human Calretinin (Calbindin 2) protein fragment (around aa23-242) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title