missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Calpain 1 Antibody (15C10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30796-100ug
This item is not returnable.
View return policy
Description
Calpain 1 Monoclonal antibody specifically detects Calpain 1 in Bovine,Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| Calpain 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS and 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| 15C10 | |
| Western Blot : 1-2 μg/mL, ELISA : 0.5-1 μg/mL, Immunoprecipitation | |
| Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with human Calpain 1 purified from placenta with mouse myeloma. | |
| 100 μg | |
| Cell Cycle and Replication | |
| 823 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction