missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Calpain 1 Antibody (15C10), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-30796-25ug

 View more versions of this product

Product Code. 30067135

  • 213.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calpain 1 Monoclonal antibody specifically detects Calpain 1 in Bovine,Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calpain 1
Monoclonal
Unconjugated
PBS and 50% glycerol
Mouse
Protein A or G purified
RUO
Primary
Bovine, Human, Mouse, Rat
Purified
ELISA, Immunoprecipitation, Western Blot
15C10
Western Blot : 1-2 μg/mL, ELISA : 0.5-1 μg/mL, Immunoprecipitation
Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL
Hybridoma produced by the fusion of splenocytes from mice immunized with human Calpain 1 purified from placenta with mouse myeloma.
25 μg
Cell Cycle and Replication
823
Store at -20°C. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.