Learn More
CALML4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15894-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| CALML4 | |
| Polyclonal | |
| Unconjugated | |
| CALML4 | |
| CALML4, calmodulin like 4, Calmodulin like protein 4, NY BR 20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 691455, 75600, 91860 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91WQ9, Q96GE6 | |
| Calml4 | |
| CALML4 Fusion Protein Ag8705 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |