missing translation for 'onlineSavingsMsg'
Learn More

California Residual Solvent Calibration Mixture 2 10000 μg/mL in Triacetin, Dr. Ehrenstorfer

Product Code. 30325042
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30325042

Brand: Dr. Ehrenstorfer DRES50000047TN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1, 75-05-8, 60-29-7, 64-17-5, 141-78-6, 67-63-0
Quantity 5 x 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Triacetin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.