Learn More
California Pesticides Class 2B Mixture 1035 100 μg/mL in Acetonitrile, Second Source, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Hexythiazox∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methomyl∣Myclobutanil∣Naled∣Oxamyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Pyrethrins∣Pyridaben∣Quintozene∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiamethoxam∣Trifloxystrobin |
| CAS | 78587-05-0, 138261-41-3, 143390-89-0, 121-75-5, 57837-19-1, 16752-77-5 |
| Analyte or Component Names | Hexythiazox∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methomyl∣Myclobutanil∣Naled∣Oxamyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Pyrethrins∣Pyridaben∣Quintozene∣Spinetoram J∣Spinosad∣Spiromesifen∣Spirotetramat∣Tebuconazole∣Thiamethoxam∣Trifloxystrobin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Hexythiazox 100 μg/mL∣Imidacloprid 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Malathion 100 μg/mL∣Metalaxyl 100 μg/mL∣Methomyl 100 μg/mL∣Myclobutanil 100 μg/mL∣Naled 100 μg/mL∣Oxamyl 100 μg/mL∣Permethrin 100 μg/mL∣Phosmet 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Prallethrin 100 μg/mL∣Propiconazole 100 μg/mL∣Pyrethrins 100 μg/mL∣Pyridaben 100 μg/mL∣Quintozene 100 μg/mL∣Spinetoram J 100 μg/mL∣Spinosad 100 μg/mL∣Spiromesifen 100 μg/mL∣Spirotetramat 100 μg/mL∣Tebuconazole 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL |