missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calicivirus Antibody (FCV1-43), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63286AF647
This item is not returnable.
View return policy
Description
Calicivirus Monoclonal specifically detects Calicivirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Calicivirus | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-63286 specifically recognizes feline calicivirus, and does not cross-react with FIV or FeLV.The antibody detects a 63kD band in Western blots, representing the viral capsid protein. It may also be used in immunofluorescence techniques to detect viral infection, and has neutralising activity against feline calicivirus. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FCV1-43 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FCV | |
| Calcivirus viral capsid protein (63 kDa) | |
| 0.125 mL | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction