missing translation for 'onlineSavingsMsg'
Learn More

CALCOCO2 Monoclonal Antibody (OTI2D11), TrueMAB™, OriGene

Product Code. 17585651
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17585651 30 μL 30µL
17565651 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17585651 Supplier Origene Technologies Supplier No. TA501972S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CALCOCO2 Monoclonal antibody specifically detects CALCOCO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen CALCOCO2
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2D11
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CALCOCO2
Gene Accession No. Q13137
Gene Alias 2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974
Host Species Mouse
Immunogen Full length human recombinant protein of human CALCOCO2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10241
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.