missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitonin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB600-1156-25ul
This item is not returnable.
View return policy
Description
Calcitonin Polyclonal antibody specifically detects Calcitonin in Human, Mouse, Rat, Gerbil, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Calcitonin | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:4000 | |
| P01257 | |
| Sheep | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Gerbil, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS | |
| ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
| Synthetic peptide corresponding to a portion of rat alpha-calcitonin gene-related peptide (CGRP). | |
| 25 μg | |
| Cancer, Cellular Markers, Neuroscience, Phospho Specific | |
| 796 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction