missing translation for 'onlineSavingsMsg'
Learn More

Calbindin D-28K Antibody (rCALB1/13280), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-45090

Product Code. 30448922

  • 593.00€ / 100µg
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calbindin D-28K
Monoclonal
1 mg/mL
Western Blot : 2-4ug/mL, Immunohistochemistry : 1-2ug/mL
CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Recombinant fragment (around aa 7-96) of human Calbindin 1/2 protein (Uniprot: P05937). Exact sequence is proprietary.
100 μg
Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
793
Store at -20 to -84°C. Avoid freeze-thaw cycles.
Purified
Immunohistochemistry, Western Blot
rCALB1/13280
Unconjugated
10 mM PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.