missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CADM3 Monoclonal Antibody (730014)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561336
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Cell adhesion molecule 3 (CADM3), also known as Necl-1, is a gene located on chromosome 1p36.22, encoding a protein that is part of the immunoglobulin superfamily involved in cell-cell adhesion processes. CADM3 is predominantly expressed in the nervous system, particularly in regions such as the brain and spinal cord, where it plays crucial roles in neuronal communication, synapse formation, and maintenance of neural network architecture. The protein functions by mediating homophilic and heterophilic interactions with other cell adhesion molecules, facilitating synapse stabilization and neuronal differentiation. CADM3's activity is essential for proper neural development and function, and dysregulation of its expression has been linked to various neurological disorders. Beyond its role in the nervous system, CADM3 also contributes to tumor suppression in certain cancers, as its expression is often downregulated in tumor tissues. Research into CADM3 continues to explore its potential as a therapeutic target in both neurobiology and oncology.
Specifications
| CADM3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q8N126 | |
| Cadm3 | |
| Mouse myeloma cell line NS0-derived recombinant human IGSF4B/SynCAM3, Pro21-Tyr329. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 730014 | |
| Unconjugated | |
| Cadm3 | |
| BIgR; brain immunoglobulin receptor; CADM3; Cell adhesion molecule 3; dendritic cell nectin-like protein 1 short isoform; IGSF4B; Immunoglobulin superfamily member 4B; immunoglobulin superfamily, member 4B; NECL1; NECL-1; nectin-like 1; nectin-like protein 1; nectin-lke 1; synaptic cell adhesion molecule 3; SYNCAM3; TSLC1-like 1; TSLC1-like protein 1; TSLL1; UNQ225/PRO258 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 57863 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction