Learn More
CACNB3 Antibody, PE, R&D Systems™
Product Code. 18600936 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18600936 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18600936 Supplier Novus Biologicals Supplier No. NBP299461PE
 Restricted Item
Estimated Shipment: 14-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
CACNB3 Polyclonal specifically detects CACNB3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen CACNB3
Classification Polyclonal
Dilution Western Blot
Formulation PBS
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNB3.
Regulatory Status RUO
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Product Line Novus