missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABYR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12351-1-AP-150UL
This item is not returnable.
View return policy
Description
Spermatozoa gain fertilization capacitation and hyperactivation after residing in the uterus or oviduct. Calcium, cAMP and protein tyrosine phosphorylation are necessary for the molecular mechanisms that allow for this transformation. CABYR (Calcium-binding tyrosine phosphorylation-regulated protein), also known as Fibrousheathin-2 or Cancer/testis antigen 88, is a 493 amino acid protein that is expressed in sperm flagella and exhibits increased tyrosine phosphorylation during capacitation. There are six named isoforms of CABYR that are produced as a result of alternative slicing events. Specifically, isoform 1 is expressed in the testis, while isoform 3 and isoform 5 are expressed in pancreas, brain and various brain tumors. Isoforms 1, 2 and 6 most likely bind calcium, whereas isoforms 3, 4 and 5 probably do not bind calcium.Specifications
| CABYR | |
| Polyclonal | |
| Unconjugated | |
| CABYR | |
| CABYR, Calcium binding protein 86, Cancer/testis antigen 88, CBP86, CT88, Fibrousheathin 2, Fibrousheathin II, FSP 2, FSP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26256, 71132 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75952, Q9D424 | |
| Cabyr | |
| CABYR Fusion Protein Ag3012 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction