missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C8orf4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26279-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a small, monomeric, predomitly unstructured protein that functions as a positive regulator of the Wnt/beta-catenin signaling pathway. This protein interacts with a repressor of beta-catenin mediated transcription at nuclear speckles. It is thought to competitively block interactions of the repressor with beta-catenin, resulting in up-regulation of beta-catenin target genes. The encoded protein may also play a role in the NF-kappaB and ERK1/2 signaling pathways. Expression of this gene may play a role in the proliferation of several types of cancer including thyroid cancer, breast cancer and hematological maligcies.Specifications
| C8orf4 | |
| Polyclonal | |
| Unconjugated | |
| C8orf4 | |
| C8orf4, hTC 1, TC 1, TC1, Thyroid cancer protein 1, Uncharacterized protein C8orf4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56892, 69068 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NR00 | |
| C8orf4, Tcim | |
| C8orf4 Fusion Protein Ag23930 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction