missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf182 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24957-1-AP-20UL
This item is not returnable.
View return policy
Description
C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.Specifications
| C6orf182 | |
| Polyclonal | |
| Unconjugated | |
| CEP57L1 | |
| C6orf182, Centrosomal protein 57kDa-like protein 1, Cep57 related protein, CEP57L1, Cep57R | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103268, 285753 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IYX8, Q8VDS7 | |
| CEP57L1 | |
| C6orf182 Fusion Protein Ag21054 | |
| 20 μL | |
| Primary | |
| Non-human Primate, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction