missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ C6orf115 Recombinant Protein

Brand:  Abnova™ H00058527-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193113

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH14953
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
34.65
8
Glutathione Sepharose 4 Fast Flow
25 ug
MNVDHEVNLLVEEIHRLGSKNADGKLSVKFGVLFRDDKCANLFEALVGTLKAAKRRKIVTYPGELLLQGVHDDVDIILLQD
HSPC280/PRO2013
C6orf115
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
58527
C6orf115 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf115
Human
Recombinant
Solution
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers