Learn More
C5b-9 Antibody (2A1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-77377-0.1MG
This item is not returnable.
View return policy
Description
C5b-9 Monoclonal specifically detects C5b-9 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.
Specifications
| C5b-9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay | |
| MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex | |
| Rat C5b-9 | |
| Store at 4°C. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2A1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| 0.1 mg | |
| Purified |