Learn More
C5b-9 Antibody (2A1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-77377-0.025MG
 View more versions of this product
Product Code. 18397710
  • 175.00€ / 25µg
Description
C5b-9 Monoclonal specifically detects C5b-9 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C5b-9
Monoclonal
0.5 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay
MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex
Rat C5b-9
Rat
Purified
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoassay, Flow Cytometry, Immunofluorescence
2A1
Unconjugated
PBS
Mouse
0.025 mg
Store at 4°C.