Learn More
C5aR Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-21316-100UL
 View more versions of this product
Product Code. 18095095
  • 450.00€ / 100µL
Description
The C5aR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C5aR. This antibody recognizes human antigen. The C5aR antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
C5aR
1000 μg/mL
C5A, C5a R, C5AR, C5AR1, C5R1, CD88
100 μL
728
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa