Learn More
C5aR Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-21316-100UL
Description
The C5aR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C5aR. This antibody recognizes human antigen. The C5aR antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| C5aR | |
| 1000 μg/mL | |
| C5A, C5a R, C5AR, C5AR1, C5R1, CD88 | |
| 100 μL | |
| 728 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |